5-(4-Dimethylaminobenzylidene)rhodanine, 99%
5-(4-Dimethylaminobenzylidene)rhodanine, 99%
5-(4-Dimethylaminobenzylidene)rhodanine, 99%
5-(4-Dimethylaminobenzylidene)rhodanine, 99%
Thermo Scientific Chemicals
5-(4-Dimethylaminobenzylidene)rhodanine, 99%
5-(4-Dimethylaminobenzylidene)rhodanine, 99%, ACROS Organics, Quantity: 100g, Packaging: Glass bottle, Red, Melting Point: 285 deg.C to 288 deg.C, Molecular Weight: 264.35, Percent Purity: 99%, Appearance: (0.3% soln. in acetone) clear, Assay Percent Range: 99%, Beilstein: 27, 433, CAS: 536-17-4
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
25 g
Packaging:
Glass bottle
Catalog number FSA151471000
Price (MYR)
2,998.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,998.00
Add to cart
Chemical Identifiers
CAS34513-98-9
IUPAC Nameλ²-ruthenium(2+) iminooxidanium trinitrate
Molecular FormulaN4O10Ru
InChI KeyWOSOOWIGVAKGOC-UHFFFAOYSA-N
SMILES[Ru++].N#[O+].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear orange to dark brown
Appearance (Form)Solution
Assay1.5 % Ru (typically)