Thymol Blue, Indicator
Thymol Blue, Indicator
Thymol Blue, Indicator
Thymol Blue, Indicator
Thermo Scientific Chemicals
Thymol Blue, Indicator
CAS: 76-61-9 | C27H30O5S | 466.592 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA151451000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS54-11-5
IUPAC Name3-[(2S)-1-methylpyrrolidin-2-yl]pyridine
Molecular FormulaC10H14N2
InChI KeySNICXCGAKADSCV-JTQLQIEISA-N
SMILESCN1CCC[C@H]1C1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.5 % (K.F.)
Appearance (Color)Clear colorless to yellow to brown
GC>=99.0 %
Specific optical rotationon anhydrous basis
View more
Indicator
RUO – Research Use Only