Phenol Red, sodium salt
Phenol Red, sodium salt
Phenol Red, sodium salt
Phenol Red, sodium salt
Thermo Scientific Chemicals
Phenol Red, sodium salt
CAS: 34487-61-1 | C19H13NaO5S | 376.358 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA151441000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS9005-65-6
Molecular Formula(C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
InChI KeyHDTIFOGXOGLRCB-KTKRTIGZNA-N
SMILESCCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
MDL NumberMFCD00082107
SpecificationsSpecification SheetSpecification Sheet
Acid Value≤2.0 mg KOH/g
Identification (FTIR)Conforms (non-U.S. sourced material)
Hydroxyl Value65.0-80.0 mg KOH/g
Appearance (Color)Yellow to orange
CommentMaterial sourced in the U.S. and in other countries
View more
Indicator
RUO – Research Use Only