Methyl orange, Indicator
Methyl orange, Indicator
Methyl orange, Indicator
Methyl orange, Indicator
Thermo Scientific Chemicals
Methyl orange, Indicator
CAS: 547-58-0 | C14H14N3NaO3S | 327.334 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA151425000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS13478-18-7
IUPAC Namemolybdenum(3+) trichloride
Molecular FormulaCl3Mo
InChI KeyZSSVQAGPXAAOPV-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Mo+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
Indicator
RUO – Research Use Only