Methyl Red, sodium salt, indicator, pure
Methyl Red, sodium salt, indicator, pure
Methyl Red, sodium salt, indicator, pure
Methyl Red, sodium salt, indicator, pure
Thermo Scientific Chemicals
Methyl Red, sodium salt, indicator, pure
CAS: 845-10-3 | C15H14N3NaO2 | 291.286 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA151411000
Price (MYR)
624.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
624.00
Add to cart
Chemical Identifiers
CAS7787-64-6
IUPAC Namebismuth(3+) triiodide
Molecular FormulaBiI3
InChI KeyKOECRLKKXSXCPB-UHFFFAOYSA-K
SMILES[I-].[I-].[I-].[Bi+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤0.001% (10 ppm)
Appearance (Color)Grey to black
FormPowder and/or granules
Assay (special)?2.5 % of Theoretical value (Bi)
Indicator
RUO – Research Use Only