Methyl Yellow, pure, indicator grade
Methyl Yellow, pure, indicator grade
Methyl Yellow, pure, indicator grade
Methyl Yellow, pure, indicator grade
Thermo Scientific Chemicals
Methyl Yellow, pure, indicator grade
CAS: 60-11-7 | C14H15N3 | 225.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA151401000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS109-55-7
IUPAC Name(3-aminopropyl)dimethylamine
Molecular FormulaC5H14N2
InChI KeyIUNMPGNGSSIWFP-UHFFFAOYSA-N
SMILESCN(C)CCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)>98.5%
Refractive Index1.4335-1.4375 @ 20?C
FormLiquid
Indicator
RUO – Research Use Only