Cresol Red, Indicator Grade, Pure
Cresol Red, Indicator Grade, Pure
Cresol Red, Indicator Grade, Pure
Cresol Red, Indicator Grade, Pure
Thermo Scientific Chemicals
Cresol Red, Indicator Grade, Pure
CAS: 1733-12-6 | C21H18O5S | 382.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA151381000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6156-78-1
IUPAC Namemanganese(2+) diacetate tetrahydrate
Molecular FormulaC4H14MnO8
InChI KeyCESXSDZNZGSWSP-UHFFFAOYSA-L
SMILESO.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light pink
Titration Complexometric>=99.0 %
Chloride (Cl)=<20 ppm
Copper (Cu)=<5 ppm
Insoluble matter=<0.01 % (in water)
View more
Indicator
RUO – Research Use Only