Bromothymol Blue, sodium salt, pure, water soluble, indicator
CAS: 34722-90-2 | C27H27Br2NaO5S | 646.37 g/mol
Have Questions?
Change view
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA151370250
Price (MYR)
570.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
570.00
Add to cart
Specifications
Chemical Name or MaterialBromothymol Blue, sodium salt
CAS34722-90-2
EINECS Number252-169-7
TSCATSCA
Molecular FormulaC27H27Br2NaO5S