Bromophenol Blue sodium salt, ACS reagent
Bromophenol Blue sodium salt, ACS reagent
Bromophenol Blue sodium salt, ACS reagent
Bromophenol Blue sodium salt, ACS reagent
Thermo Scientific Chemicals
Bromophenol Blue sodium salt, ACS reagent
Indicator
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number FSA151351000
Price (MYR)
1,053.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
1,053.00
Add to cart
Chemical Identifiers
CAS9042-11-9
MDL NumberMFCD00132693
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Particle size=<10 % (< 75 µm)
Appearance (Color)White to off-white
Total weight capacity>=10 meq/g
Particle size=<10 % (> 150 µm)
View more
Indicator
RUO – Research Use Only