Bromophenol Blue, sodium salt, pure, water soluble, indicator
Bromophenol Blue, sodium salt, pure, water soluble, indicator
Bromophenol Blue, sodium salt, pure, water soluble, indicator
Bromophenol Blue, sodium salt, pure, water soluble, indicator
Thermo Scientific Chemicals
Bromophenol Blue, sodium salt, pure, water soluble, indicator
CAS: 62625-28-9 | C19H9Br4NaO5S | 691.942 g/mol
Have Questions?
Catalog NumberQuantity
FSA15135025025 g
Catalog number FSA151350250
Price (MYR)
470.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
470.00
Add to cart
Chemical Identifiers
CAS603-85-0
IUPAC Name2-amino-3-nitrophenol
Molecular FormulaC6H6N2O3
InChI KeyKUCWUAFNGCMZDB-UHFFFAOYSA-N
SMILESNC1=C(O)C=CC=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red
FormPowder
Assay (GC)≥97.5%
Indicator
RUO – Research Use Only