Bromophenol Blue, sodium salt, pure, water soluble, indicator
Bromophenol Blue, sodium salt, pure, water soluble, indicator
Bromophenol Blue, sodium salt, pure, water soluble, indicator
Bromophenol Blue, sodium salt, pure, water soluble, indicator
Thermo Scientific Chemicals
Bromophenol Blue, sodium salt, pure, water soluble, indicator
CAS: 62625-28-9 | C19H9Br4NaO5S | 691.942 g/mol
Have Questions?
Catalog NumberQuantity
FSA1513500505 g
Catalog number FSA151350050
Price (MYR)
212.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
212.00
Add to cart
Chemical Identifiers
CAS622-26-4
IUPAC Name4-(2-hydroxyethyl)piperidin-1-ium
Molecular FormulaC7H16NO
InChI KeyLDSQQXKSEFZAPE-UHFFFAOYSA-O
SMILESOCCC1CC[NH2+]CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point45°C to 50°C
Appearance (Color)White
Appearance (Form)Low melting crystalline solid
GC>=96.0 %
Infrared spectrumConforms
Indicator
RUO – Research Use Only