Bromophenol Blue, Indicator, Pure
Bromophenol Blue, Indicator, Pure
Bromophenol Blue, Indicator, Pure
Bromophenol Blue, Indicator, Pure
Thermo Scientific Chemicals
Bromophenol Blue, Indicator, Pure
Bromophenol Blue, C19H10Br4O5S, CAS Number-115-39-9, 3', 3″, 5', 3',3″,5',5″-Tetrabromophenolsulfonephthalein, BPB, 25g, 19, I, 649, CHEBI:59424, Orange-Pink to Red, 669.96, Pure, UDSAIICHUKSCKT-UHFFFAOYSA-N, Authentic
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA151340250
Price (MYR)
556.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
556.00
Add to cart
Specifications
Chemical Name or MaterialBromophenol Blue
Melting Point279.0°C
Name NoteIndicator
CAS115-39-9
Molecular FormulaC19H10Br4O5S
View more
Indicator
RUO – Research Use Only