Bromocresol Purple, pure, indicator grade
Bromocresol Purple, pure, indicator grade
Bromocresol Purple, pure, indicator grade
Bromocresol Purple, pure, indicator grade
Thermo Scientific Chemicals
Bromocresol Purple, pure, indicator grade
CAS: 115-40-2 | C21H16Br2O5S | 540.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
140 g
5 g
25 g
1 kg
Packaging:
Glass bottle
Catalog number FSA151331400
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
140 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS565-61-7
IUPAC Name3-methylpentan-2-one
Molecular FormulaC6H12O
InChI KeyUIHCLUNTQKBZGK-UHFFFAOYNA-N
SMILESCCC(C)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless
GC>=98.5 %
Appearance (Form)Liquid
Indicator
RUO – Research Use Only