Bromocresol Green, Water Soluble, Indicator
Bromocresol Green, Water Soluble, Indicator
Bromocresol Green, Water Soluble, Indicator
Bromocresol Green, Water Soluble, Indicator
Thermo Scientific Chemicals
Bromocresol Green, Water Soluble, Indicator
CAS: 67763-24-0 | C21H13Br4NaO5S | 720.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA151320250
Price (MYR)
607.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
607.00
Add to cart
Chemical Identifiers
CAS137-40-6
IUPAC Namesodium propanoate
Molecular FormulaC3H5NaO2
InChI KeyJXKPEJDQGNYQSM-UHFFFAOYSA-M
SMILES[Na+].CCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HClO499.0 to 100.5 % (On dried substance)
Water=<1 % (K.F.)
Heavy metals (as Pb)=<10 ppm
View more
Indicator
RUO – Research Use Only