Bromocresol Green, pure, indicator grade
Bromocresol Green, pure, indicator grade
Bromocresol Green, pure, indicator grade
Bromocresol Green, pure, indicator grade
Thermo Scientific Chemicals
Bromocresol Green, pure, indicator grade
Bromocresol Green, C21H14Br4O5S, CAS Number-76-60-8, 3', 3″, 5', 3',3″,5',5″-Tetrabromo-m-cresolsulfonephthalein, BCG, 5g, Brown, 200-972-8, 698.01, Pure, Warning, GHS H Statement: May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation.
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA151310250
Price (MYR)
819.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
819.00
Add to cart
Chemical Identifiers
CAS107-21-1
IUPAC Nameethane-1,2-diol
Molecular FormulaC2H6O2
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILESOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear viscous liquid
Infrared spectrumConforms
Sulfated ash=<0.005 %
Iron (Fe)=<0.5 ppm
Water=<0.1 % (K.F.)
View more
Indicator
RUO – Research Use Only