Isopropyl acetate, 99%, pure, Thermo Scientific Chemicals
Isopropyl acetate, 99%, pure, Thermo Scientific Chemicals
Isopropyl acetate, 99%, pure, Thermo Scientific Chemicals
Isopropyl acetate, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isopropyl acetate, 99%, pure, Thermo Scientific Chemicals
CAS: 108-21-4 | C5H10O2 | 102.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number FSA150860010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialIsopropyl acetate
Name Note99%
CAS108-21-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.