tert-Butylhydroquinone, 97%, Thermo Scientific Chemicals
tert-Butylhydroquinone, 97%, Thermo Scientific Chemicals
tert-Butylhydroquinone, 97%, Thermo Scientific Chemicals
tert-Butylhydroquinone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
tert-Butylhydroquinone, 97%, Thermo Scientific Chemicals
CAS: 1948-33-0 | C10H14O2 | 166.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
500 g
Packaging:
View all
Plastic bottle
Catalog number FSA150822500
Price (MYR)
369.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
369.00
Add to cart
Chemical Identifiers
CAS7697-37-2
IUPAC Namenitric acid
Molecular FormulaHNO3
InChI KeyGRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILESO[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofABarium(Ba)
Appearance (Color)Colorless to yellow, ≤10 APHA
Assay from Supplier's CofA67.0 - 70.0%
Assay from Supplier's CofAAluminum (Al)
Assay from Supplier's CofAAntimony (Sb)