tert-Butylhydroquinone, 97%, Thermo Scientific Chemicals
tert-Butylhydroquinone, 97%, Thermo Scientific Chemicals
tert-Butylhydroquinone, 97%, Thermo Scientific Chemicals
tert-Butylhydroquinone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
tert-Butylhydroquinone, 97%, Thermo Scientific Chemicals
CAS: 1948-33-0 | C10H14O2 | 166.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
500 g
Packaging:
View all
Glass Bottle
Catalog number FSA150820050
Price (MYR)
226.00
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
226.00
Add to cart
Specifications
Chemical Name or Materialtert-Butylhydroquinone
Name Note97%
CAS1948-33-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Very toxic to aquatic life with long lasting effects.