1-Butanethiol, 98%, Thermo Scientific Chemicals
1-Butanethiol, 98%, Thermo Scientific Chemicals
1-Butanethiol, 98%, Thermo Scientific Chemicals
1-Butanethiol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Butanethiol, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number FSA150800010
Price (MYR)
938.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
938.00
Add to cart
Chemical Identifiers
CAS76058-33-8
IUPAC Name2-{2-[4-(diethylamino)phenyl]diazen-1-yl}benzoate
Molecular FormulaC17H18N3O2
InChI KeyHBRCDTRQDHMTDA-UHFFFAOYSA-M
SMILESCCN(CC)C1=CC=C(C=C1)N=NC1=CC=CC=C1C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark red to dark purple powder
FormPowder
Color ChangeRed @ pH 4.5 to yellow @ pH 6.5
lambda-max (UV-Vis)450nm ± 5nm (0.1M NaOH)
Solution Test0.1% in ethanol: clear