N,N-Diethylethylenediamine, 98+%, Thermo Scientific Chemicals
N,N-Diethylethylenediamine, 98+%, Thermo Scientific Chemicals
N,N-Diethylethylenediamine, 98+%, Thermo Scientific Chemicals
N,N-Diethylethylenediamine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Diethylethylenediamine, 98+%, Thermo Scientific Chemicals
CAS: 100-36-7 | C6H16N2 | 116.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA150705000
Price (MYR)
824.00
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
824.00
Add to cart
Specifications
Chemical Name or MaterialN, N-Diethylethylenediamine
Name Note98+%
CAS100-36-7
CAS Min %98.0
Health Hazard 1GHS Signal Word: Danger