Formaldehyde Sodium Bisulfite Addition Compound 95%, Thermo Scientific Chemicals
Formaldehyde Sodium Bisulfite Addition Compound 95%, Thermo Scientific Chemicals
Formaldehyde Sodium Bisulfite Addition Compound 95%, Thermo Scientific Chemicals
Formaldehyde Sodium Bisulfite Addition Compound 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Formaldehyde Sodium Bisulfite Addition Compound 95%, Thermo Scientific Chemicals
CAS: 870-72-4 | CH3NaO4S | 134.081 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
5 g
1 kg
Packaging:
View all
Plastic Drum
Catalog number FSA150690051
Price (MYR)
2,088.00
Add to cart
Quantity:
5 kg
Packaging:
Plastic Drum
Request bulk or custom format
Price (MYR)
2,088.00
Add to cart
Chemical Identifiers
CAS13400-13-0
IUPAC Namecaesium(1+) fluoride
Molecular FormulaCsF
InChI KeyXJHCXCQVJFPJIK-UHFFFAOYSA-M
SMILES[F-].[Cs+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay>=98.5 %
Potassium (K)=<300 ppm
Strontium (Sr)=<10 ppm
Aluminium (Al)=<10 ppm