Ethyl formate, 98+%, pure, Thermo Scientific Chemicals
Ethyl formate, 98+%, pure, Thermo Scientific Chemicals
Ethyl formate, 98+%, pure, Thermo Scientific Chemicals
Ethyl formate, 98+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethyl formate, 98+%, pure, Thermo Scientific Chemicals
CAS: 109-94-4 | C3H6O2 | 74.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Packaging:
Glass bottle
Catalog number FSA150675000
Price (MYR)
399.00
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
399.00
Add to cart
Chemical Identifiers
CAS16069-36-6
IUPAC Name2,5,8,15,18,21-hexaoxatricyclo[20.4.0.0⁹,¹⁴]hexacosane
Molecular FormulaC20H36O6
InChI KeyBBGKDYHZQOSNMU-UHFFFAOYNA-N
SMILESC1CCC2OCCOCCOC3CCCCC3OCCOCCOC2C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥96.0%