Isoamyl acetate, 99%, pure, mixture of 2- and 3-Methylbutyl acetate, Thermo Scientific Chemicals
Isoamyl acetate, 99%, pure, mixture of 2- and 3-Methylbutyl acetate, Thermo Scientific Chemicals
Isoamyl acetate, 99%, pure, mixture of 2- and 3-Methylbutyl acetate, Thermo Scientific Chemicals
Isoamyl acetate, 99%, pure, mixture of 2- and 3-Methylbutyl acetate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isoamyl acetate, 99%, pure, mixture of 2- and 3-Methylbutyl acetate, Thermo Scientific Chemicals
CAS: 123-92-2 | C7H14O2 | 130.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number FSA150660010
Price (MYR)
562.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
562.00
Add to cart
Chemical Identifiers
CAS67-56-1
IUPAC Namemethanol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-UHFFFAOYSA-N
SMILESCO
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.005 % (Coulometric)
GC>=99.8 %
Acidity/alkalinity=<0.0005 meq/g
Appearance (Form)Clear liquid