4-Butylaniline, 97%, Thermo Scientific Chemicals
4-Butylaniline, 97%, Thermo Scientific Chemicals
4-Butylaniline, 97%, Thermo Scientific Chemicals
4-Butylaniline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Butylaniline, 97%, Thermo Scientific Chemicals
CAS: 104-13-2 | C10H15N | 149.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA150651000
Price (MYR)
496.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
496.00
Add to cart
Specifications
Chemical Name or Material4-Butylaniline
CAS104-13-2
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0