Chemical Identifiers
CAS13268-42-3
IUPAC Nametriammonium hexaoxo-1,4,6,9,10,13-hexaoxa-5-ferraspiro[4.4⁵.4⁵]tridecane-5,5,5-tris(ylium) trihydrate
Molecular FormulaC6H18FeN3O15
InChI KeyARJSTCBPHIPXIJ-UHFFFAOYSA-K
SMILES[NH4+].[NH4+].[NH4+].O.O.O.O=C1O[Fe+3]234(OC1=O)OC(=O)C(=O)O2.O=C(O3)C(=O)O4
View more