L(-)-Malic acid, 99%, Thermo Scientific Chemicals
L(-)-Malic acid, 99%, Thermo Scientific Chemicals
L(-)-Malic acid, 99%, Thermo Scientific Chemicals
L(-)-Malic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L(-)-Malic acid, 99%, Thermo Scientific Chemicals
CAS: 97-67-6 | C4H6O5 | 134.087 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Plastic bottle
Catalog number FSA150591000
Price (MYR)
425.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
425.00
Add to cart
Specifications
Chemical Name or MaterialL(-)-Malic acid
CAS97-67-6
Melting Point99°C to 107°C
Health Hazard 1Danger