L(-)-Malic acid, 99%, Thermo Scientific Chemicals
L(-)-Malic acid, 99%, Thermo Scientific Chemicals
L(-)-Malic acid, 99%, Thermo Scientific Chemicals
L(-)-Malic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L(-)-Malic acid, 99%, Thermo Scientific Chemicals
CAS: 97-67-6 | C4H6O5 | 134.087 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number FSA150590250
Price (MYR)
259.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
259.00
Add to cart
Chemical Identifiers
CAS429-41-4
IUPAC Nametetrabutylazanium fluoride
Molecular FormulaC16H36FN
InChI KeyFPGGTKZVZWFYPV-UHFFFAOYSA-M
SMILES[F-].CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Water=<8 %
Appearance (Color)Clear yellow to amber to light green to brown
Titration with HClO40.90 to 1.10 M