Ethyl propionate, 99+%, Thermo Scientific Chemicals
Ethyl propionate, 99+%, Thermo Scientific Chemicals
Ethyl propionate, 99+%, Thermo Scientific Chemicals
Ethyl propionate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethyl propionate, 99+%, Thermo Scientific Chemicals
CAS: 105-37-3 | C5H10O2 | 102.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
Packaging:
Glass bottle
Catalog number FSA150505000
Price (MYR)
318.00
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
318.00
Add to cart
Specifications
Chemical Name or MaterialEthyl propionate
Acidity1mg KOH/g max.
CAS105-37-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Highly flammable liquid and vapor.