4-Nitrobenzoyl chloride, 98%, Thermo Scientific Chemicals
4-Nitrobenzoyl chloride, 98%, Thermo Scientific Chemicals
4-Nitrobenzoyl chloride, 98%, Thermo Scientific Chemicals
4-Nitrobenzoyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Nitrobenzoyl chloride, 98%, Thermo Scientific Chemicals
CAS: 122-04-3 | C7H4ClNO3 | 185.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
250 g
Packaging:
Glass bottle
Catalog number FSA150435000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1936-15-8
IUPAC Namedisodium (8Z)-7-oxo-8-(2-phenylhydrazin-1-ylidene)-7,8-dihydronaphthalene-1,3-disulfonate
Molecular FormulaC16H10N2Na2O7S2
InChI KeyAEOVEGJBKQQFOP-DDVLFWKVSA-L
SMILES[Na+].[Na+].[O-]S(=O)(=O)C1=CC2=C(\C(=N\NC3=CC=CC=C3)C(=O)C=C2)C(=C1)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
UV spectrumComplies
Appearance (Color)Orange to orange-red
Dye content>=85.0 %