4-Bromobenzyl bromide, 98%, Thermo Scientific Chemicals
4-Bromobenzyl bromide, 98%, Thermo Scientific Chemicals
4-Bromobenzyl bromide, 98%, Thermo Scientific Chemicals
4-Bromobenzyl bromide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Bromobenzyl bromide, 98%, Thermo Scientific Chemicals
CAS: 589-15-1 | C7H6Br2 | 249.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA150410250
Price (MYR)
335.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
335.00
Add to cart
Chemical Identifiers
CAS29676-71-9
IUPAC Name2-(2-amino-1,3-thiazol-4-yl)acetic acid
Molecular FormulaC5H6N2O2S
InChI KeyDYCLHZPOADTVKK-UHFFFAOYSA-N
SMILESNC1=NC(CC(O)=O)=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Appearance (Color)White to pale cream