2,3-Butanedione monoxime, 98%, Thermo Scientific Chemicals
2,3-Butanedione monoxime, 98%, Thermo Scientific Chemicals
2,3-Butanedione monoxime, 98%, Thermo Scientific Chemicals
2,3-Butanedione monoxime, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,3-Butanedione monoxime, 98%, Thermo Scientific Chemicals
CAS: 57-71-6 | C4H7NO2 | 101.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA150371000
Price (MYR)
580.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
580.00
Add to cart
Chemical Identifiers
CAS17745-45-8
IUPAC Namepropylboronic acid
Molecular FormulaC3H9BO2
InChI KeyJAQOMSTTXPGKTN-UHFFFAOYSA-N
SMILESCCCB(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5%
Melting Point (clear melt)104.0-111.0?C
Proton NMRConforms to structure