2,3-Butanedione monoxime, 98%, Thermo Scientific Chemicals
2,3-Butanedione monoxime, 98%, Thermo Scientific Chemicals
2,3-Butanedione monoxime, 98%, Thermo Scientific Chemicals
2,3-Butanedione monoxime, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,3-Butanedione monoxime, 98%, Thermo Scientific Chemicals
CAS: 57-71-6 | C4H7NO2 | 101.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Plastic bottle
Catalog number FSA150370025
Price (MYR)
7,340.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
7,340.00
Add to cart
Chemical Identifiers
CAS2978-58-7
IUPAC Name2-methylbut-3-yn-2-amine
Molecular FormulaC5H9N
InChI KeyVUGCBIWQHSRQBZ-UHFFFAOYSA-N
SMILESCC(C)(N)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow
FormLiquid
Assay (GC)>94.0%