Hippuric acid, 98%, Thermo Scientific Chemicals
Hippuric acid, 98%, Thermo Scientific Chemicals
Hippuric acid, 98%, Thermo Scientific Chemicals
Hippuric acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hippuric acid, 98%, Thermo Scientific Chemicals
CAS: 495-69-2 | C9H9NO3 | 179.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
Packaging:
View all
Plastic bottle
Catalog number FSA150271000
Price (MYR)
262.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
262.00
Add to cart
Specifications
Chemical Name or MaterialHippuric acid
Name Note98%
CAS495-69-2
Health Hazard 1GHS Signal Word: Danger
Melting Point188.0°C to 191.0°C