Hippuric acid, 98%, Thermo Scientific Chemicals
Hippuric acid, 98%, Thermo Scientific Chemicals
Hippuric acid, 98%, Thermo Scientific Chemicals
Hippuric acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hippuric acid, 98%, Thermo Scientific Chemicals
CAS: 495-69-2 | C9H9NO3 | 179.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Glass Bottle
Catalog number FSA150270050
Price (MYR)
230.00
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
230.00
Add to cart
Chemical Identifiers
CAS80747-90-6
SpecificationsSpecification SheetSpecification Sheet
Particle Size<300 µm: ≤1.0%
Particle Size>1180 µm: ≤5.0%
Particle SizeParticle Diameter: 500-750 µm
Assay (special)Uniformity Coefficient: ≤1.80
Assay (special)Total Exchange Capacity: 1.60-1.71 eq/L