Ethyl 4-hydroxybenzoate, 99%, Thermo Scientific Chemicals
Ethyl 4-hydroxybenzoate, 99%, Thermo Scientific Chemicals
Ethyl 4-hydroxybenzoate, 99%, Thermo Scientific Chemicals
Ethyl 4-hydroxybenzoate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethyl 4-hydroxybenzoate, 99%, Thermo Scientific Chemicals
CAS: 120-47-8 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
Catalog number FSA150251000
Price (MYR)
270.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
270.00
Add to cart
Chemical Identifiers
CAS78922-04-0
IUPAC Name3-(3-chlorobenzenesulfonamido)benzoic acid
Molecular FormulaC13H10ClNO4S
InChI KeyAPBOVLPLJFJSRI-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(NS(=O)(=O)C2=CC(Cl)=CC=C2)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormSpherical beads
Appearance (Color)Pale brown to brown
CommentMax reversible swelling Na -> H ca. 10%
Exchange capacity≥2.00 eq/L (Na+ form)
MatrixStyrene divinylbenzene copolymer