Ethyl 4-hydroxybenzoate, 99%, Thermo Scientific Chemicals
Ethyl 4-hydroxybenzoate, 99%, Thermo Scientific Chemicals
Ethyl 4-hydroxybenzoate, 99%, Thermo Scientific Chemicals
Ethyl 4-hydroxybenzoate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethyl 4-hydroxybenzoate, 99%, Thermo Scientific Chemicals
CAS: 120-47-8 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
250 g
Catalog number FSA150250010
Price (MYR)
830.00
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
830.00
Add to cart
Chemical Identifiers
CAS999-78-0
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥97.0% (GC, U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
Refractive Index1.4050-1.4100 @ 20°C (non-U.S. specification)
Appearance (Color)Clear colorless to pale yellow