n-Propyl Ether, 99+%
n-Propyl Ether, 99+%
n-Propyl Ether, 99+%
n-Propyl Ether, 99+%
n-Propyl Ether, 99+%
CAS: 111-43-3 | C6H14O | 102.18 g/mol
Have Questions?
Catalog number FSA150040500
Price (MYR)
992.00
Add to cart
Request bulk or custom format
Price (MYR)
992.00
Add to cart
Chemical Identifiers
CAS22068-49-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)114.0-120.0?C