trans-2-Hexene, 98+%, Thermo Scientific Chemicals
trans-2-Hexene, 98+%, Thermo Scientific Chemicals
trans-2-Hexene, 98+%, Thermo Scientific Chemicals
trans-2-Hexene, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
trans-2-Hexene, 98+%, Thermo Scientific Chemicals
CAS: 4050-45-7 | C6H12 | 84.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
Packaging:
Glass bottle
Catalog number FSA149900050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7785-23-1
IUPAC Namesilver(1+) bromide
Molecular FormulaAgBr
InChI KeyADZWSOLPGZMUMY-UHFFFAOYSA-M
SMILES[Br-].[Ag+]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Yellow/green
Assay (special)99.5-100.0%