4-Heptanol, 93%
4-Heptanol, 93%
4-Heptanol, 93%
Thermo Scientific Chemicals
4-Heptanol, 93%
CAS: 589-55-9 | C7H16O | 116.2 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA1498500505 mLGlass bottle
Catalog number FSA149850050
Price (MYR)
407.00
Add to cart
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
407.00
Add to cart
Specifications
Chemical Name or Material4-Heptanol
Name Note93%
CAS589-55-9
CAS Min %91.0
Health Hazard 1GHS Signal Word: Warning