Valeric Acid, 99%
Valeric Acid, 99%
Valeric Acid, 99%
Thermo Scientific Chemicals
Valeric Acid, 99%
CAS: 109-52-4 | C5H10O2 | 102.133 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
5 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA149570025
Price (MYR)
921.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
921.00
Add to cart
Chemical Identifiers
CAS108-12-3
IUPAC Name3-methylbutanoyl chloride
Molecular FormulaC5H9ClO
InChI KeyISULZYQDGYXDFW-UHFFFAOYSA-N
SMILESCC(C)CC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.4135-1.4175 @ 20?C
Appearance (Color)Clear colorless