Acetonitrile, 99+%, extra pure, Thermo Scientific Chemicals
Acetonitrile, 99+%, extra pure, Thermo Scientific Chemicals
Acetonitrile, 99+%, extra pure, Thermo Scientific Chemicals
Acetonitrile, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetonitrile, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 75-05-8 | C2H3N | 41.053 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA149520025
Price (MYR)
1,265.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,265.00
Add to cart
Specifications
Chemical Name or MaterialAcetonitrile
Melting Point-46.0°C
CAS75-05-8
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Highly flammable liquid and vapor.