Acetonitrile, 99+%, extra pure, Thermo Scientific Chemicals
Acetonitrile, 99+%, extra pure, Thermo Scientific Chemicals
Acetonitrile, 99+%, extra pure, Thermo Scientific Chemicals
Acetonitrile, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetonitrile, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 75-05-8 | C2H3N | 41.053 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number FSA149520010
Price (MYR)
540.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
540.00
Add to cart
Chemical Identifiers
CAS142-82-5
IUPAC Nameheptane
Molecular FormulaC7H16
InChI KeyIMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILESCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=99.0 %
Appearance (Color)Clear colorless
Infrared spectrumConforms
Residue after evaporation=<20 ppm