1-Propanol, 99+%, Extra Pure, Thermo Scientific Chemicals
1-Propanol, 99+%, Extra Pure, Thermo Scientific Chemicals
1-Propanol, 99+%, Extra Pure, Thermo Scientific Chemicals
1-Propanol, 99+%, Extra Pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Propanol, 99+%, Extra Pure, Thermo Scientific Chemicals
CAS: 71-23-8 | C3H8O | 60.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number FSA149480010
Price (MYR)
404.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
404.00
Add to cart
Chemical Identifiers
CAS13494-80-9
IUPAC Nametellurium
Molecular FormulaTe
InChI KeyPORWMNRCUJJQNO-UHFFFAOYSA-N
SMILES[Te]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.