Phenol, 99%, extra pure, Thermo Scientific Chemicals
Phenol, 99%, extra pure, Thermo Scientific Chemicals
Phenol, 99%, extra pure, Thermo Scientific Chemicals
Phenol, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenol, 99%, extra pure, Thermo Scientific Chemicals
CAS: 108-95-2 | C6H6O | 94.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
50 g
5 kg
Packaging:
View all
Glass bottle
Catalog number FSA149340010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1600-27-7
IUPAC Namemercury(2+) diacetate
Molecular FormulaC4H6HgO4
InChI KeyBRMYZIKAHFEUFJ-UHFFFAOYSA-L
SMILES[Hg++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration Complexometric>=97.5 %
Sulfate (SO4)=<100 ppm
Appearance (Color)White to almost white
Melting point179°C to 182°C