Methacrylamide, 98%, extra pure
Methacrylamide, 98%, extra pure
Methacrylamide, 98%, extra pure
Methacrylamide, 98%, extra pure
Thermo Scientific Chemicals
Methacrylamide, 98%, extra pure
CAS: 79-39-0 | C4H7NO | 85.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Catalog number FSA149205000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS327-97-9
IUPAC Name(1S,3R,4R,5R)-3-{[(2E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy}-1,4,5-trihydroxycyclohexane-1-carboxylic acid
Molecular FormulaC16H18O9
InChI KeyCWVRJTMFETXNAD-JUHZACGLSA-N
SMILESO[C@@H]1C[C@](O)(C[C@@H](OC(=O)\C=C\C2=CC=C(O)C(O)=C2)[C@@H]1O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
Appearance (Color)White to yellow to light beige
HPLC>=97.5 %