Xanthine, 98%, Thermo Scientific Chemicals
Xanthine, 98%, Thermo Scientific Chemicals
Xanthine, 98%, Thermo Scientific Chemicals
Xanthine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Xanthine, 98%, Thermo Scientific Chemicals
CAS: 69-89-6 | C5H4N4O2 | 152.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA149170050
Price (MYR)
338.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
338.00
Add to cart
Chemical Identifiers
CAS10034-76-1
IUPAC Namedicalcium hydrate disulfate
Molecular FormulaCa2H2O9S2
InChI KeyZOMBKNNSYQHRCA-UHFFFAOYSA-J
SMILESO.[Ca++].[Ca++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration Complexometric>=97.0 % (On dry substance)
Loss on ignition4.5 % to 8.0 % (500°C, 2 h)
Appearance (Color)White to off-white
IdentificationConfirmed