Iodoform, 99+%, Thermo Scientific Chemicals
Iodoform, 99+%, Thermo Scientific Chemicals
Iodoform, 99+%, Thermo Scientific Chemicals
Iodoform, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iodoform, 99+%, Thermo Scientific Chemicals
CAS: 75-47-8 | CHI3 | 393.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number FSA149121000
Price (MYR)
513.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
513.00
Add to cart
Chemical Identifiers
CAS68938-92-1
IUPAC Nametetrabromoplatinumtetrakis(ylium)
Molecular FormulaBr4Pt
InChI KeyDYFRBYUUSCNRBP-UHFFFAOYSA-J
SMILESBr[Pt+4](Br)(Br)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.