Aminoguanidine bicarbonate, 98.5%, Thermo Scientific Chemicals
Aminoguanidine bicarbonate, 98.5%, Thermo Scientific Chemicals
Aminoguanidine bicarbonate, 98.5%, Thermo Scientific Chemicals
Aminoguanidine bicarbonate, 98.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Aminoguanidine bicarbonate, 98.5%, Thermo Scientific Chemicals
CAS: 2582-30-1 | CH6N4·H2CO3 | 136.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Plastic Bottle
Catalog number FSA149062500
Price (MYR)
262.00
Add to cart
Quantity:
250 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
262.00
Add to cart
Chemical Identifiers
CAS6226-79-5
IUPAC Nametetrasodium (4Z)-3-oxo-4-(2-{2-sulfonato-5-[2-(4-sulfonatophenyl)diazen-1-yl]phenyl}hydrazin-1-ylidene)-3,4-dihydronaphthalene-2,7-disulfonate
Molecular FormulaC22H12N4Na4O13S4
InChI KeyLZYGEJPERQQCIY-STNZDNLRSA-J
SMILES[Na+].[Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)N=NC1=CC=C(C(N\N=C2/C(=O)C(=CC3=CC(=CC=C23)S([O-])(=O)=O)S([O-])(=O)=O)=C1)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Loss on Drying≤5.0%
Assay (Dye)Dye Content: ≥80.0%
Solution TestConforms (0.1% Water)
Appearance (Color)Red-brown to dark brown