Benzoic Acid, Sodium Salt, 99+%, Thermo Scientific Chemicals
Benzoic Acid, Sodium Salt, 99+%, Thermo Scientific Chemicals
Benzoic Acid, Sodium Salt, 99+%, Thermo Scientific Chemicals
Benzoic Acid, Sodium Salt, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzoic Acid, Sodium Salt, 99+%, Thermo Scientific Chemicals
CAS: 532-32-1 | C7H5NaO2 | 144.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
View all
Plastic Bottle
Catalog number FSA148980010
Price (MYR)
305.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
305.00
Add to cart
Specifications
Chemical Name or MaterialSodium benzoate
CAS532-32-1
CAS Min %99.0
CAS Max %100.0