3,3'-Dithiodipropionic acid, 98%, Thermo Scientific Chemicals
3,3'-Dithiodipropionic acid, 98%, Thermo Scientific Chemicals
3,3'-Dithiodipropionic acid, 98%, Thermo Scientific Chemicals
3,3'-Dithiodipropionic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,3'-Dithiodipropionic acid, 98%, Thermo Scientific Chemicals
CAS: 1119-62-6 | C6H10O4S2 | 210.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Glass bottle
Catalog number FSA148900500
Price (MYR)
281.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
281.00
Add to cart
Chemical Identifiers
CAS531-75-9
IUPAC Name7-hydroxy-6-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2H-chromen-2-one
Molecular FormulaC15H16O9
InChI KeyXHCADAYNFIFUHF-TYKRLAFXNA-N
SMILESOC[C@H]1O[C@@H](OC2=C(O)C=C3OC(=O)C=CC3=C2)[C@H](O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation(on dry substance)
Infrared spectrumConforms
Titration with NaOH>=96.0 % (On dry substance)
Specific optical rotation-84° to -87° (20°C, 589 nm) (c=1.5, dioxane/H2O 1:1)
Appearance (Color)White to cream